SY3H30995107 Synthonix Corporation

4-Bromo-1H-benzo[d]imidazol-2-amine

  • MDL number No MDL found
  • Cas Number : 1266114-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/32443.html

Safety Information

MSDS https://synthonix.com/32443.html
Availability details for
Quantity
  • Availability
  • Add to Cart