SY3H30995108 Synthonix Corporation

6-Bromo-4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD16618981
  • Cas Number : 1203684-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32447.html

Safety Information

MSDS https://synthonix.com/32447.html
Availability details for
Quantity
  • Availability
  • Add to Cart