SY3H30995109 Synthonix Corporation

4-Bromo-1,2-dihydropyridazine-3,6-dione

  • MDL number MFCD12923098
  • Cas Number : 15456-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/32458.html

Safety Information

MSDS https://synthonix.com/32458.html
Availability details for
Quantity
  • Availability
  • Add to Cart