SY3H3099510C Synthonix Corporation

6-((tert-butyldimethylsilyl)oxy)hexan-1-amine

  • Cas Number : 124883-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/8554.html

Safety Information

MSDS https://synthonix.com/8554.html
Availability details for
Quantity
  • Availability
  • Add to Cart