SY3H30995112 Synthonix Corporation

2-Chloro-4-methyl-6-(trifluoromethyl)pyridine

  • MDL number MFCD10700262
  • Cas Number : 749256-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/32253.html

Safety Information

MSDS https://synthonix.com/32253.html
Availability details for
Quantity
  • Availability
  • Add to Cart