SY3H30995118 Synthonix Corporation

6-Chloro-3-iodoimidazo[1,2-a]pyrazine

  • MDL number MFCD18072709
  • Cas Number : 1245645-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/32310.html

Safety Information

MSDS https://synthonix.com/32310.html
Availability details for
Quantity
  • Availability
  • Add to Cart