SY3H3099511E Synthonix Corporation

1-(2-Chloropyrimidin-4-yl)ethanone

  • MDL number MFCD22543973
  • Cas Number : 1312535-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/32407.html

Safety Information

MSDS https://synthonix.com/32407.html
Availability details for
Quantity
  • Availability
  • Add to Cart