SY3H30995122 Synthonix Corporation

4-Chloro-7-(trifluoromethyl)quinazoline

  • MDL number MFCD09837196
  • Cas Number : 16499-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/32451.html

Safety Information

MSDS https://synthonix.com/32451.html
Availability details for
Quantity
  • Availability
  • Add to Cart