SY3H3099512A Synthonix Corporation

3,5-Dichloro-2-iodopyrazine

  • MDL number No MDL found
  • Cas Number : 136866-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/32285.html

Safety Information

MSDS https://synthonix.com/32285.html
Availability details for
Quantity
  • Availability
  • Add to Cart