SY3H315B0DEF Synthonix Corporation

2-Amino-3-ethoxypyrazine

  • MDL number MFCD08460065
  • Cas Number : 89464-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36915.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36915.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart