SY3H315B0E05 Synthonix Corporation

(3-Amino-6-bromopyrazin-2-yl)methanol

  • MDL number MFCD13193331
  • Cas Number : 1260880-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37107.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37107.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart