SY3H315B0E0F Synthonix Corporation

5-Amino-2,3-difluorobenzonitrile

  • MDL number MFCD20726994
  • Cas Number : 1247885-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Gray to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37208.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37208.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart