SY3H315B0E16 Synthonix Corporation

5-Acetonyl-2-methoxybenzene sulfonamide

  • MDL number MFCD07782136
  • Cas Number : 116091-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: Very pale yellow - Pale reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37371.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37371.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart