SY3H315B0E54 Synthonix Corporation

5-Bromo-1,3-dihydrobenzoimidazol-2-one

  • MDL number MFCD01893032
  • Cas Number : 39513-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36673.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36673.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart