SY3H315B0E73 Synthonix Corporation

4-Bromo-2-vinylpyridine

  • MDL number MFCD09833154
  • Cas Number : 502509-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36792.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36792.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart