SY3H315B0E91 Synthonix Corporation

2-Bromo-5-(trifluoromethyl)isonicotinic acid

  • MDL number MFCD16610250
  • Cas Number : 823222-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36919.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36919.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart