SY3H315B0ED9 Synthonix Corporation

8-Bromo-4-chloroquinazoline

  • MDL number MFCD09738589
  • Cas Number : 125096-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37198.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37198.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart