SY3H315B0EDE Synthonix Corporation

1-Bromo-2-chloro-6-iodobenzene

  • MDL number MFCD28397823
  • Cas Number : 1369793-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37218.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37218.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart