SY3H315B0EEC Synthonix Corporation

2-(3-Bromopyridin-2-yl)propan-2-ol

  • MDL number MFCD16988539
  • Cas Number : 1240594-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37253.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37253.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart