SY3H315B0F01 Synthonix Corporation

7-Bromoimidazo[1,2-a]pyridine-3-carboxylic acid

  • MDL number MFCD09994702
  • Cas Number : 1019021-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37325.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37325.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart