SY3H315B0F15 Synthonix Corporation

Benzyltrimethylammonium dichloroiodate

  • MDL number MFCD00075259
  • Cas Number : 114971-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: Pale yellow - Reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37400.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37400.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart