SY3H315B0F32 Synthonix Corporation

2-Bromo-3-chloropyrazine

  • MDL number MFCD14581973
  • Cas Number : 1206250-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Off White to light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37498.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37498.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart