SY3H315B0F4D Synthonix Corporation

[2,2'-Bipyridin]-3-amine

  • MDL number MFCD13180435
  • Cas Number : 105166-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Off white to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37603.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/37603.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart