SY3H315B0F52 Synthonix Corporation

4-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one

  • MDL number MFCD12963348
  • Cas Number : 1190318-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37615.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/37615.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart