SY3H315B0F62 Synthonix Corporation

4-Bromo-5-chloro-2-fluorobenzoic acid

  • MDL number MFCD19686184
  • Cas Number : 1349708-91-3
  • Molecular Weight : 253.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-5-chloro-2-fluorobenzoic acid
Inchi Code 1S/C7H3BrClFO2/c8-4-2-6(10)3(7(11)12)1-5(4)9/h1-2H,(H,11,12)
Inchi Key FQYCZAHBPRQWRR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to off-white to pale-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/37657.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37657.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart