SY3H315B0F73 Synthonix Corporation

3-Bromo-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride

  • MDL number MFCD09259954
  • Cas Number : 1172057-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37732.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37732.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart