SY3H315B0F74 Synthonix Corporation

3-Bromo-4,5-dichloropyridine

  • MDL number MFCD17014545
  • Cas Number : 1001056-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37738.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37738.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart