SY3H315B0FC0 Synthonix Corporation

5-Cyclopropylpyridin-2-ol

  • MDL number MFCD12033218
  • Cas Number : 1159821-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37180.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37180.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart