SY3H315B0FD8 Synthonix Corporation

5-Chloro-3-(trifluoromethyl)pyridin-2-ol

  • MDL number MFCD13185452
  • Cas Number : 1214342-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37398.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37398.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart