SY3H315B1019 Synthonix Corporation

Dimethyl 4-chloropyridine-2,6-dicarboxylate

  • MDL number MFCD02612781
  • Cas Number : 5371-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Off white to yellow to very pale reddish yellow to brown or pale-red powder or crytals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36623.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36623.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart