SY3H315B1024 Synthonix Corporation

2,5-Dibromo-4-fluoroaniline

  • MDL number MFCD18089317
  • Cas Number : 172377-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Yellow to Brown or Gray-green Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36710.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36710.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart