SY3H315B102B Synthonix Corporation

2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD00051958
  • Cas Number : 4425-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36777.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36777.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart