SY3H315B1033 Synthonix Corporation

2,3-Dihydro-1H-pyrrolo[2,3-c]pyridine

  • MDL number MFCD09907942
  • Cas Number : 760919-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36872.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36872.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart