SY3H315B1037 Synthonix Corporation

Diphenyl(2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphine

  • MDL number MFCD26961066
  • Cas Number : 819867-23-7
  • Molecular Weight : 464.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diphenyl(2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl)phosphane
Inchi Code 1S/C33H37P/c1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28/h7-25H,1-6H3
Inchi Key HCULUUWFPUUYEE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/36886.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/36886.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart