SY3H315B107B Synthonix Corporation

2,6-Difluoro-4-methylbenzaldehyde

  • MDL number MFCD11847293
  • Cas Number : 1201597-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37538.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37538.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart