SY3H315B108D Synthonix Corporation

2,8-Dibromodibenzo[b,d]furan

  • MDL number MFCD00093683
  • Cas Number : 10016-52-1
  • Molecular Weight : 325.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,8-dibromodibenzo[b,d]furan
Inchi Code 1S/C12H6Br2O/c13-7-1-3-11-9(5-7)10-6-8(14)2-4-12(10)15-11/h1-6H
Inchi Key UFCZRCPQBWIXTR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/37733.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37733.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart