SY3H315B1092 Synthonix Corporation

ethyl 5-cyano-1H-indole-2-carboxylate

  • MDL number MFCD04966999
  • Cas Number : 105191-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/16275.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/16275.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart