SY3H315B1093 Synthonix Corporation

Ethyl 6-aminopicolinate

  • MDL number MFCD01009842
  • Cas Number : 69142-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/32237.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/32237.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart