SY3H315B1098 Synthonix Corporation

Ethyl 4-cyanopiperidine-4-carboxylate hydrochloride

  • MDL number MFCD24387236
  • Cas Number : 123730-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/32324.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/32324.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart