SY3H315B10A0 Synthonix Corporation

Ethyl 5-bromo-1-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD17168482
  • Cas Number : 1269293-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/32430.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/32430.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart