SY3H315B10AE Synthonix Corporation

5-Ethoxypyridin-3-amine

  • MDL number MFCD01646192
  • Cas Number : 51468-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/36780.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/36780.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart