SY3H315B10C8 Synthonix Corporation

Ethyl 5-(hydroxymethyl)isoxazole-3-carboxylate

  • MDL number MFCD08273501
  • Cas Number : 123770-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37262.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37262.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart