SY3H315B10CE Synthonix Corporation

Ethyl 4-chlorophenylacetate

  • MDL number MFCD00018791
  • Cas Number : 14062-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Colorless or White to light yellow liquid or low-melting solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37347.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37347.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart