SY3H315B110D Synthonix Corporation

4-Fluoro-5-iodo-1H-indole

  • MDL number MFCD22628143
  • Cas Number : 1240113-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37256.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37256.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart