SY3H315B1113 Synthonix Corporation

2-Fluoro-3-nitrobenzonitrile

  • MDL number MFCD11849938
  • Cas Number : 1214328-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to brown or yellow-green solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37405.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37405.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart