SY3H315B111B Synthonix Corporation

5-Fluoro-1H-pyrrolo[2,3-c]pyridine

  • MDL number MFCD12962571
  • Cas Number : 1190319-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white powder or crystal
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37610.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37610.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart