SY3H315B1125 Synthonix Corporation

tert-butyl 4-formylpyridin-3-ylcarbamate

  • MDL number MFCD02683209
  • Cas Number : 116026-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/9439.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/9439.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart