SY3H315B1135 Synthonix Corporation

4-Hydroxy-2-(trifluoromethoxy)benzaldehyde

  • MDL number MFCD06797920
  • Cas Number : 1017083-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37005.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37005.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart