SY3H315B1137 Synthonix Corporation

1-(3-Hydroxypyridin-2-yl)ethanone

  • MDL number MFCD08062669
  • Cas Number : 13210-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37046.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37046.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart