SY3H315B114A Synthonix Corporation

2-Hydrazinylbenzonitrile hydrochloride

  • MDL number MFCD03094641
  • Cas Number : 1030287-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Yellow to yellow-green powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/37662.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/37662.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart